Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159714.1 | NP_001153186.1 | 11 - 12 | 1213 | 63 | |
NM_027328.4 | NP_081604.3 | 11 - 12 | 1231 | 63 | ||
GenBank mRNA | AK033190.1 | - | 11 - 12 | 1234 | 63 | |
AK033283.1 | - | 11 - 12 | 1196 | 63 | ||
AK051260.1 | - | 11 - 12 | 1191 | 63 | ||
AY040823.1 | - | 11 - 12 | 1207 | 63 | ||
BC018376.1 | - | 11 - 12 | 1197 | 3673893 | 63 | |
BC057877.1 | - | 10 - 11 | 1174 | 5347609 | 63 | |
BC061461.1 | - | 11 - 12 | 1157 | 5717606 | 63 | |
CT010189.1 | - | 10 - 11 | 1151 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |