Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008290.2 | NP_032316.2 | 2 - 3 | 529 | 67 | |
GenBank mRNA | AK004950.1 | - | 2 - 3 | 530 | 67 | |
BC012682.1 | - | 2 - 3 | 518 | 4217153 | 67 | |
BE649625.1 | - | 1 - 2 | 160 | 67 | ||
X95685.1 | - | 2 - 3 | 490 | 67 | ||
Y09517.1 | - | 2 - 3 | 573 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |