Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024243.4 | NP_077205.3 | 5 - 6 | 999 | 62 | |
GenBank mRNA | AK002230.1 | - | 5 - 6 | 967 | 62 | |
AK053323.1 | - | 5 - 6 | 944 | 62 | ||
AK089958.1 | - | 5 - 6 | 969 | 62 | ||
AK150526.1 | - | 5 - 6 | 934 | 62 | ||
AK150890.1 | - | 5 - 6 | 989 | 62 | ||
AK151258.1 | - | 5 - 6 | 967 | 62 | ||
AK151361.1 | - | 5 - 6 | 988 | 62 | ||
AK151879.1 | - | 5 - 6 | 988 | 62 | ||
AK151908.1 | - | 5 - 6 | 942 | 62 | ||
AK152070.1 | - | 5 - 6 | 942 | 62 | ||
AK165604.1 | - | 5 - 6 | 969 | 62 | ||
AK170503.1 | - | 5 - 6 | 979 | 62 | ||
BC003235.1 | - | 5 - 6 | 972 | 3497631 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |