Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163689.1 | NP_001157161.1 | 7 - 8 | 1153 | 72 | |
NM_025802.3 | NP_080078.2 | 6 - 7 | 985 | 72 | ||
NR_028142.1 | 8 - 9 | 1235 | 72 | |||
GenBank mRNA | AK002826.1 | - | 6 - 7 | 967 | 72 | |
AK003207.1 | - | 2 - 3 | 207 | 72 | ||
AK031609.1 | - | 7 - 8 | 1111 | 72 | ||
AK076464.1 | - | 7 - 8 | 937 | 72 | ||
AK085926.1 | - | 7 - 8 | 1138 | 72 | ||
AY510273.1 | - | 7 - 8 | 1134 | 72 | ||
AY731699.1 | - | 7 - 8 | 1134 | 72 | ||
AY894805.1 | - | 7 - 8 | 1054 | 72 | ||
BC019188.1 | - | 8 - 9 | 1217 | 5027544 | 72 | |
BC034190.1 | - | 6 - 7 | 654 | 72 | ||
BC044737.1 | - | 8 - 9 | 1199 | 72 | ||
BC044781.2 | - | 8 - 9 | 1199 | 5151911 | 72 | |
BC054825.1 | - | 8 - 9 | 1115 | 72 | ||
BC064747.1 | - | 7 - 8 | 1125 | 30024535 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |