Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003604.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003604.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003604.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003601.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003601.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003601.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003609.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003609.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003609.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003605.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003605.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003605.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003606.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003606.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003606.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003603.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003603.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003603.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003602.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003602.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003602.mm.1 |
NM_029555.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003608.mm.1 |
TC0600000454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003608.mm.1 |
NM_029555.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003608.mm.1 |
NM_029555.2 |
Refseq |