Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133810.3 | NP_598571.2 | 3 - 4 | 522 | 83 | |
GenBank mRNA | AK029551.1 | - | 3 - 4 | 463 | 83 | |
AK035693.1 | - | 3 - 4 | 490 | 83 | ||
AK076932.1 | - | 3 - 4 | 489 | 83 | ||
AK083332.1 | - | 3 - 4 | 491 | 83 | ||
AK153108.1 | - | 3 - 4 | 466 | 83 | ||
AK165291.1 | - | 3 - 4 | 462 | 83 | ||
AY092028.1 | - | 3 - 4 | 475 | 83 | ||
BC006579.1 | - | 3 - 4 | 477 | 3586767 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |