Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133879.2 | NP_598640.2 | 4 - 5 | 1156 | 69 | |
GenBank mRNA | AM161454.1 | - | 4 - 5 | 1104 | 69 | |
BC005515.1 | - | 4 - 5 | 1167 | 3493611 | 69 | |
CX202255.1 | - | 2 - 3 | 333 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |