Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133978.2 | NP_598739.1 | 3 - 4 | 600 | 68 | |
GenBank mRNA | AF188504.1 | - | 3 - 4 | 436 | 68 | |
AK151785.1 | - | 3 - 4 | 403 | 68 | ||
AK167886.1 | - | 3 - 4 | 446 | 68 | ||
AK168024.1 | - | 3 - 4 | 453 | 68 | ||
AK168280.1 | - | 3 - 4 | 437 | 68 | ||
AY241869.1 | - | 3 - 4 | 457 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |