Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026550.3 | NP_080826.2 | 3 - 4 | 689 | 65 | |
GenBank mRNA | AF386076.1 | - | 3 - 4 | 675 | 65 | |
AK002852.1 | - | 3 - 4 | 439 | 65 | ||
AK077846.1 | - | 3 - 4 | 677 | 65 | ||
AK077941.1 | - | 3 - 4 | 436 | 65 | ||
AK135946.1 | - | 3 - 4 | 436 | 65 | ||
AK150747.1 | - | 3 - 4 | 437 | 65 | ||
AK150919.1 | - | 3 - 4 | 437 | 65 | ||
AK188749.1 | - | 2 - 3 | 191 | 65 | ||
AY030406.1 | - | 3 - 4 | 504 | 65 | ||
BC018522.1 | - | 3 - 4 | 432 | 65 | ||
BC060371.1 | - | 3 - 4 | 433 | 65 | ||
BC116729.1 | - | 3 - 4 | 420 | 65 | ||
BC116731.1 | - | 3 - 4 | 420 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |