Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285931.1 | NP_001272860.1 | 8 - 9 | 2763 | 78 | |
NM_001285932.1 | NP_001272861.1 | 8 - 9 | 2529 | 78 | ||
NM_027883.3 | NP_082159.3 | 8 - 9 | 2254 | 78 | ||
GenBank mRNA | AK004733.1 | - | 8 - 9 | 2255 | 78 | |
AK036763.1 | - | 4 - 5 | 655 | 78 | ||
AK076217.1 | - | 7 - 8 | 2230 | 78 | ||
AK129062.1 | - | 8 - 9 | 2237 | 78 | ||
BC058370.1 | - | 8 - 9 | 2591 | 78 | ||
BC080856.1 | - | 8 - 9 | 2529 | 6854931 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |