Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028057.2 | NP_082333.1 | 1 - 2 | 70 | 84 | |
GenBank mRNA | AK005159.1 | - | 1 - 2 | 71 | 84 | |
AK159663.1 | - | 1 - 2 | 40 | 84 | ||
AK167178.1 | - | 1 - 2 | 64 | 84 | ||
BC016266.1 | - | 1 - 2 | 279 | 4924006 | 84 | |
BC024618.1 | - | 1 - 2 | 48 | 4235599 | 84 | |
JX420289.1 | - | 1 - 2 | 505 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |