Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134078.4 | NP_598839.2 | 3 - 4 | 729 | 71 | |
GenBank mRNA | AK211372.1 | - | 1 - 2 | 75 | 71 | |
BC024115.1 | - | 3 - 4 | 717 | 5100379 | 71 | |
BC033365.1 | - | 3 - 4 | 700 | 5367653 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |