Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145779.1 | NP_001139251.1 | 17 - 18 | 1832 | 131 | |
NM_008442.2 | NP_032468.2 | 19 - 20 | 2329 | 131 | ||
GenBank mRNA | AK037037.1 | - | 17 - 18 | 1745 | 131 | |
BB622253.1 | - | 1 - 2 | 102 | 131 | ||
BC006803.1 | - | 17 - 18 | 1827 | 3152883 | 131 | |
BE863830.1 | - | 5 - 6 | 502 | 131 | ||
D12644.1 | - | 19 - 20 | 2330 | 131 | ||
Y15894.1 | - | 16 - 17 | 1682 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |