Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009759.4 | NP_033889.2 | 4 - 5 | 487 | 85 | |
GenBank mRNA | AF012104.1 | - | 3 - 4 | 340 | 85 | |
AK036707.1 | - | 3 - 4 | 346 | 85 | ||
AK040936.1 | - | 4 - 5 | 488 | 85 | ||
AK080038.1 | - | 4 - 5 | 488 | 85 | ||
BC138104.1 | - | 4 - 5 | 364 | 8861123 | 85 | |
BC138105.1 | - | 4 - 5 | 364 | 8861124 | 85 | |
U88091.1 | - | 4 - 5 | 473 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |