Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Klhl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025727.3 | NP_080003.1 | 2 - 3 | 792 | 129 | |
GenBank mRNA | AK014906.1 | - | 2 - 3 | 793 | 129 | |
AK077202.1 | - | 2 - 3 | 794 | 129 | ||
AY495337.1 | - | 2 - 3 | 793 | 129 | ||
BC139267.1 | - | 2 - 3 | 768 | 8862289 | 129 | |
BC139268.1 | - | 2 - 3 | 768 | 8862290 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |