Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D730048I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026593.3 | NP_080869.1 | 1 - 2 | 144 | 65 | |
GenBank mRNA | AK020299.1 | - | 1 - 2 | 116 | 65 | |
AK020302.1 | - | 1 - 2 | 116 | 65 | ||
AK020310.1 | - | 1 - 2 | 97 | 65 | ||
AK021351.1 | - | 1 - 2 | 116 | 65 | ||
AK033813.1 | - | 1 - 2 | 145 | 65 | ||
AK136605.1 | - | 134 | 65 | |||
AK136636.1 | - | 1 - 2 | 115 | 65 | ||
BC027498.1 | - | 1 - 2 | 78 | 3465762 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |