Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030260.3 | NP_084536.2 | 2 - 3 | 1232 | 74 | |
NM_173002.3 | NP_766590.1 | 2 - 3 | 1232 | 74 | ||
GenBank mRNA | AK030287.1 | - | 2 - 3 | 1148 | 74 | |
AK042190.1 | - | 2 - 3 | 1159 | 74 | ||
AK042392.1 | - | 2 - 3 | 1142 | 74 | ||
AK044429.1 | - | 2 - 3 | 1125 | 74 | ||
AK044529.1 | - | 2 - 3 | 1156 | 74 | ||
AK044948.1 | - | 2 - 3 | 1126 | 74 | ||
AK047079.1 | - | 2 - 3 | 1114 | 74 | ||
AK047538.1 | - | 2 - 3 | 1148 | 74 | ||
AK080307.1 | - | 2 - 3 | 1126 | 74 | ||
AK081005.1 | - | 2 - 3 | 1234 | 74 | ||
AK169517.1 | - | 2 - 3 | 1119 | 74 | ||
AK185703.1 | - | 1 - 2 | 145 | 74 | ||
AK218430.1 | - | 1 - 2 | 145 | 74 | ||
BC003332.1 | - | 2 - 3 | 1216 | 3256958 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |