Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133994.3 | NP_598755.1 | 1 - 2 | 573 | 67 | |
GenBank mRNA | AF508157.1 | - | 1 - 2 | 573 | 67 | |
AK138669.1 | - | 1 - 2 | 230 | 67 | ||
AK154161.1 | - | 1 - 2 | 279 | 67 | ||
AK168226.1 | - | 1 - 2 | 222 | 67 | ||
BC003903.1 | - | 1 - 2 | 201 | 3601446 | 67 | |
BC057964.1 | - | 1 - 2 | 528 | 5352325 | 67 | |
BC062201.1 | - | 1 - 2 | 530 | 4913425 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |