Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hkdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145419.1 | NP_663394.1 | 5 - 6 | 684 | 85 | |
GenBank mRNA | BC016235.1 | - | 5 - 6 | 684 | 4504302 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |