Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023061.2 | NP_075548.2 | 10 - 11 | 1329 | 88 | |
GenBank mRNA | AB035508.1 | - | 10 - 11 | 1316 | 88 | |
AB035509.1 | - | 10 - 11 | 1316 | 88 | ||
BC026985.1 | - | 10 - 11 | 1316 | 5372497 | 88 | |
CJ133016.1 | - | 3 - 4 | 408 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |