Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146084.1 | NP_666196.1 | 10 - 11 | 1592 | 105 | |
GenBank mRNA | AK029766.1 | - | 11 - 12 | 1708 | 105 | |
BC031465.1 | - | 10 - 11 | 1592 | 3981146 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |