Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172304.3 | NP_758508.2 | 14 - 15 | 2867 | 111 | |
GenBank mRNA | AK132870.1 | - | 14 - 15 | 2868 | 111 | |
AK141288.1 | - | 13 - 14 | 2646 | 111 | ||
BC006867.1 | - | 11 - 12 | 2114 | 111 | ||
BC151176.1 | - | 13 - 14 | 2618 | 9088065 | 111 | |
BC151177.1 | - | 13 - 14 | 2618 | 9088066 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |