Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040426.3 | NP_001035516.2 | 14 - 15 | 2695 | 60 | |
NM_172444.4 | NP_766032.1 | 8 - 9 | 1503 | 60 | ||
GenBank mRNA | AK045414.1 | - | 8 - 9 | 1419 | 60 | |
AK138976.1 | - | 14 - 15 | 2696 | 60 | ||
BC139329.1 | - | 8 - 9 | 1396 | 8862351 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |