Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028628.1 | - | 12 - 13 | 1761 | 87 | |
AK155359.1 | - | 13 - 14 | 2474 | 87 | ||
AK162978.1 | - | 11 - 12 | 1380 | 87 | ||
AK163449.1 | - | 12 - 13 | 1743 | 87 | ||
BC025942.1 | - | 1716 | 5132457 | 87 | ||
BC031565.1 | - | 12 - 13 | 1757 | 87 | ||
BC066796.1 | - | 11 - 12 | 1704 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |