Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172310.2 | NP_758514.2 | 7 - 8 | 1003 | 89 | |
GenBank mRNA | AK040927.1 | - | 7 - 8 | 976 | 89 | |
AK044290.1 | - | 7 - 8 | 1004 | 89 | ||
AK079416.1 | - | 7 - 8 | 985 | 89 | ||
BC039225.1 | - | 7 - 8 | 987 | 4458085 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |