Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172477.4 | NP_766065.1 | 5 - 6 | 1817 | 70 | |
GenBank mRNA | AK048041.1 | - | 5 - 6 | 1802 | 70 | |
AK081176.1 | - | 5 - 6 | 1818 | 70 | ||
AK135781.1 | - | 3 - 4 | 362 | 70 | ||
AK160784.1 | - | 5 - 6 | 1579 | 70 | ||
AK165202.1 | - | 5 - 6 | 1673 | 70 | ||
AK167298.1 | - | 5 - 6 | 1671 | 70 | ||
BC056367.1 | - | 5 - 6 | 1687 | 70 | ||
BC057310.1 | - | 5 - 6 | 1687 | 70 | ||
BC145599.1 | - | 4 - 5 | 1496 | 9054165 | 70 | |
BC150799.1 | - | 4 - 5 | 1496 | 9087710 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |