Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172824.3 | NP_766412.2 | 6 - 7 | 749 | 68 | |
GenBank mRNA | AK029725.1 | - | 6 - 7 | 750 | 68 | |
AK090294.1 | - | 6 - 7 | 656 | 68 | ||
AK172014.1 | - | 1 - 2 | 1003 | 68 | ||
BC095999.1 | - | 6 - 7 | 645 | 68 | ||
CF742918.1 | - | 4 - 5 | 482 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |