Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172862.3 | NP_766450.2 | 21 - 22 | 8904 | 66 | |
GenBank mRNA | AB236662.1 | - | 21 - 22 | 8904 | 66 | |
AJ833643.1 | - | 21 - 22 | 8633 | 66 | ||
AK031494.1 | - | 11 - 12 | 1806 | 66 | ||
AK082256.1 | - | 4 - 5 | 934 | 66 | ||
BC046388.1 | - | 1 - 2 | 66 | |||
BC068185.1 | - | 11 - 12 | 1806 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |