Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172922.3 | NP_766510.1 | 5 - 6 | 908 | 65 | |
GenBank mRNA | AK036872.1 | - | 5 - 6 | 909 | 65 | |
BC145077.1 | - | 5 - 6 | 935 | 65 | ||
BC145078.1 | - | 5 - 6 | 898 | 9053626 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |