Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mb21d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173386.5 | NP_775562.2 | 2 - 3 | 998 | 113 | |
GenBank mRNA | AK030292.1 | - | 2 - 3 | 999 | 113 | |
AK054330.1 | - | 2 - 3 | 932 | 113 | ||
AK078662.1 | - | 2 - 3 | 909 | 113 | ||
AK145268.1 | - | 2 - 3 | 938 | 113 | ||
AK154165.1 | - | 2 - 3 | 967 | 113 | ||
BC052196.1 | - | 2 - 3 | 908 | 6510706 | 113 | |
BC145651.1 | - | 2 - 3 | 968 | 40130954 | 113 | |
BC145653.1 | - | 2 - 3 | 968 | 40130956 | 113 | |
KC294567.1 | - | 2 - 3 | 839 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |