Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Hexb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010422.2 | NP_034552.1 | 4 - 5 | 590 | 137 | |
GenBank mRNA | AK151149.1 | - | 4 - 5 | 527 | 137 | |
AK159134.1 | - | 4 - 5 | 543 | 137 | ||
AK159178.1 | - | 4 - 5 | 564 | 137 | ||
AK160560.1 | - | 4 - 5 | 559 | 137 | ||
AK168825.1 | - | 4 - 5 | 564 | 137 | ||
U07633.1 | - | 4 - 5 | 562 | 137 | ||
Y00964.1 | - | 4 - 5 | 503 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |