Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024291.3 | NP_077253.3 | 6 - 7 | 487 | 73 | |
GenBank mRNA | AJ293727.2 | - | 6 - 7 | 509 | 73 | |
AK047076.1 | - | 6 - 7 | 878 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |