Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252530.1 | NP_001239459.1 | 5 - 6 | 1005 | 68 | |
NM_001252531.1 | NP_001239460.1 | 5 - 6 | 948 | 68 | ||
NM_175316.3 | NP_780525.1 | 5 - 6 | 751 | 68 | ||
GenBank mRNA | AK041144.1 | - | 5 - 6 | 947 | 68 | |
AK048120.1 | - | 5 - 6 | 752 | 68 | ||
AK132394.1 | - | 5 - 6 | 1006 | 68 | ||
AK163542.1 | - | 5 - 6 | 1001 | 68 | ||
AK173049.1 | - | 3 - 4 | 537 | 68 | ||
BC019209.1 | - | 5 - 6 | 734 | 68 | ||
BC037089.1 | - | 5 - 6 | 723 | 68 | ||
BC049251.1 | - | 5 - 6 | 714 | 68 | ||
BC096485.1 | - | 5 - 6 | 760 | 5101188 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |