Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175325.3 | NP_780534.1 | 5 - 6 | 407 | 76 | |
GenBank mRNA | AK039560.1 | - | 5 - 6 | 356 | 76 | |
AK089970.1 | - | 5 - 6 | 362 | 76 | ||
AK134734.1 | - | 5 - 6 | 360 | 76 | ||
BC055797.1 | - | 5 - 6 | 327 | 76 | ||
BC089507.1 | - | 5 - 6 | 341 | 76 | ||
BC092531.1 | - | 5 - 6 | 351 | 76 | ||
BC145771.1 | - | 5 - 6 | 369 | 40131074 | 76 | |
BC145773.1 | - | 5 - 6 | 369 | 40131076 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |