Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290625.1 | NP_001277554.1 | 9 - 10 | 1349 | 90 | |
NM_001290626.1 | NP_001277555.1 | 8 - 9 | 1347 | 90 | ||
NM_177345.4 | NP_796319.1 | 9 - 10 | 1675 | 90 | ||
GenBank mRNA | AK027932.1 | - | 4 - 5 | 448 | 90 | |
AK042154.1 | - | 8 - 9 | 1337 | 90 | ||
AK052045.1 | - | 9 - 10 | 1343 | 90 | ||
AK132263.1 | - | 9 - 10 | 1339 | 90 | ||
BC027377.1 | - | 4 - 5 | 477 | 90 | ||
BC031579.1 | - | 4 - 5 | 477 | 90 | ||
BC043296.1 | - | 9 - 10 | 1574 | 90 | ||
BC090644.1 | - | 8 - 9 | 1249 | 30629263 | 90 | |
BC096618.1 | - | 9 - 10 | 1648 | 6392317 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |