Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171000.2 | NP_001164471.2 | 10 - 11 | 1396 | 90 | |
NM_001171001.1 | NP_001164472.1 | 10 - 11 | 1116 | 90 | ||
NM_021414.6 | NP_067389.5 | 10 - 11 | 1399 | 90 | ||
GenBank mRNA | AK014539.1 | - | 10 - 11 | 1347 | 90 | |
AK053527.1 | - | 10 - 11 | 1080 | 90 | ||
AK122382.1 | - | 8 - 9 | 1102 | 90 | ||
BC059255.1 | - | 6 - 7 | 571 | 90 | ||
BC079660.1 | - | 10 - 11 | 1309 | 6853550 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |