Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175155.4 | NP_780364.3 | 16 - 17 | 2286 | 71 | |
GenBank mRNA | AJ507736.1 | - | 16 - 17 | 2101 | 71 | |
AK039638.1 | - | 4 - 5 | 625 | 71 | ||
AK129216.1 | - | 11 - 12 | 1620 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |