Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177922.2 | NP_808590.1 | 8 - 9 | 873 | 105 | |
GenBank mRNA | AK153712.1 | - | 2 - 3 | 298 | 105 | |
BC048082.1 | - | 8 - 9 | 873 | 6312082 | 105 | |
BC053920.1 | - | 2 - 3 | 634 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |