Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190376.1 | NP_001177305.1 | 4 - 5 | 485 | 89 | |
NM_001190378.1 | NP_001177307.1 | 4 - 5 | 485 | 89 | ||
NM_001190379.1 | NP_001177308.1 | 4 - 5 | 485 | 89 | ||
NM_178116.4 | NP_835217.1 | 4 - 5 | 485 | 89 | ||
GenBank mRNA | AK039670.1 | - | 4 - 5 | 312 | 89 | |
AK122400.1 | - | 2 - 3 | 141 | 89 | ||
AK150186.1 | - | 4 - 5 | 337 | 89 | ||
AK169118.1 | - | 4 - 5 | 324 | 89 | ||
BC049133.1 | - | 4 - 5 | 314 | 6827800 | 89 | |
BC056395.1 | - | 4 - 5 | 314 | 6852349 | 89 | |
BC059065.1 | - | 3 - 4 | 213 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |