Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ext1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010162.2 | NP_034292.2 | 7 - 8 | 2451 | 68 | |
GenBank mRNA | AK132327.1 | - | 7 - 8 | 2428 | 68 | |
BC004741.1 | - | 7 - 8 | 1815 | 3499743 | 68 | |
U78539.1 | - | 7 - 8 | 1955 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |