Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028385.1 | NP_082661.1 | 5 - 6 | 575 | 116 | |
GenBank mRNA | AK122551.1 | - | 5 - 6 | 575 | 116 | |
BC056502.1 | - | 7 - 8 | 1111 | 116 | ||
BC083184.1 | - | 4 - 5 | 456 | 30536831 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |