Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178367.4 | NP_848144.3 | 10 - 11 | 1752 | 105 | |
GenBank mRNA | AK035177.1 | - | 10 - 11 | 1752 | 105 | |
BC011309.1 | - | 3 - 4 | 414 | 105 | ||
BC027842.1 | - | 10 - 11 | 1749 | 105 | ||
BC052172.1 | - | 10 - 11 | 1746 | 6515901 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |