Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178899.5 | NP_849230.1 | 3 - 4 | 741 | 118 | |
GenBank mRNA | AK033728.1 | - | 3 - 4 | 743 | 118 | |
AK050501.1 | - | 3 - 4 | 742 | 118 | ||
BC096374.1 | - | 3 - 4 | 728 | 4483627 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |