Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009483.1 | NP_033509.1 | 9 - 10 | 1095 | 113 | |
GenBank mRNA | AJ002730.1 | - | 8 - 9 | 554 | 113 | |
AK080370.1 | - | 9 - 10 | 1114 | 113 | ||
AK144654.1 | - | 8 - 9 | 1066 | 113 | ||
BC053433.1 | - | 9 - 10 | 1095 | 6516368 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |