Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010273.4 | NP_034403.1 | 2 - 3 | 538 | 108 | |
GenBank mRNA | AF251042.1 | - | 2 - 3 | 207 | 108 | |
AK028880.1 | - | 2 - 3 | 295 | 108 | ||
AK032869.1 | - | 2 - 3 | 284 | 108 | ||
AK036969.1 | - | 2 - 3 | 297 | 108 | ||
AK156224.1 | - | 2 - 3 | 306 | 108 | ||
AK171216.1 | - | 2 - 3 | 295 | 108 | ||
AK187541.1 | - | 1 - 2 | 65 | 108 | ||
AK209882.1 | - | 2 - 3 | 187 | 108 | ||
BC013758.1 | - | 2 - 3 | 219 | 4501084 | 108 | |
BC025606.1 | - | 2 - 3 | 307 | 108 | ||
BC037598.1 | - | 2 - 3 | 272 | 5249271 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |