Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111096.1 | NP_001104566.1 | 4 - 5 | 540 | 109 | |
NM_010747.2 | NP_034877.2 | 4 - 5 | 477 | 109 | ||
GenBank mRNA | AK028112.1 | - | 4 - 5 | 473 | 109 | |
AK152199.1 | - | 4 - 5 | 538 | 109 | ||
AK153038.1 | - | 4 - 5 | 474 | 109 | ||
AK170561.1 | - | 4 - 5 | 538 | 109 | ||
BC003253.1 | - | 4 - 5 | 440 | 3500419 | 109 | |
BC031547.1 | - | 4 - 5 | 3589207 | 109 | ||
BC157998.1 | - | 4 - 5 | 309 | 9007404 | 109 | |
BY743506.1 | - | 4 - 5 | 545 | 109 | ||
M57696.1 | - | 4 - 5 | 376 | 109 | ||
M57697.1 | - | 4 - 5 | 313 | 109 | ||
M64608.1 | - | 4 - 5 | 526 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |