Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181849.2 | NP_862897.1 | 4 - 5 | 721 | 63 | |
GenBank mRNA | AK011920.1 | - | 4 - 5 | 737 | 63 | |
AK146960.1 | - | 4 - 5 | 741 | 63 | ||
AK168626.1 | - | 4 - 5 | 735 | 63 | ||
BC031715.1 | - | 4 - 5 | 721 | 4195310 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |