Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205173.1 | NP_001192102.1 | 6 - 7 | 1437 | 67 | |
NM_183148.3 | NP_898971.1 | 9 - 10 | 1057 | 67 | ||
GenBank mRNA | BC027170.1 | - | 5 - 6 | 583 | 4952810 | 67 |
BC100381.1 | - | 9 - 10 | 1057 | 67 | ||
CN718972.1 | - | 3 - 4 | 401 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |