Limited time savings – buy 3, only pay for 2

Shop now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm00834288_m1
            See other XLR GE Assays ›
            Gene Symbol:
            Xlr 3830403N18Rik
            Gene Name:
            X-linked lymphocyte-regulated
            RIKEN cDNA 3830403N18 gene
            Gene Aliases:
            -
            Xlrl
            Chromosome Location:
            Chr.X: 53777118 - 53797716 on Build GRCm38
            Chromosome Location:
            Chr.X: 55839252 - 56153496 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            -
            Assay ID Mm00834288_m1
            Size
            Availability Inventoried
            Catalog # 4331182
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Gene Symbol: Xlr
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_001291747.1 NP_001278676.1 4 - 5 248 196
            NM_001291748.1 NP_001278677.1 4 - 5 256 196
            NM_011725.4 NP_035855.1 4 - 5 296 196
            GenBank mRNA AA623498.1 - 4 - 5 210 196
            AK012549.1 - 4 - 5 287 196
            AK035025.1 - 4 - 5 258 196
            AK035341.1 - 4 - 5 265 196
            AK088930.1 - 4 - 5 200 196
            BC005560.1 - 4 - 5 283 3965533 196
            X07967.1 - 4 - 5 231 196
            Gene Symbol: 3830403N18Rik
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            GenBank mRNA AK014417.1 - 3 - 4 276 196

            Back To Top

            More Information


            Additional Information:

            Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

            Set Membership:

            Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene Xlr
            Click here to find siRNAs for Gene 3830403N18Rik

            Gene Ontology Categories:

            Function(s) Process(es)

            spermatid development
            biological_process
            molecular_function

            MicroArray Probes:

            Agilent Affymetrix ABI Illumina

            Array Name Probe IDs Targets Target Type
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009940 AK014417.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009940 AK035341.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009940 AK088930.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009940 AK035025.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009940 BC005560.1 mouse_transcriptome
            Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124964 - -
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009940 AK014417.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009940 AK035341.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009940 AK088930.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009940 AK035025.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009940 BC005560.1 mouse_transcriptome
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32389093 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09295830 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09295858 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124964 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09297577 - -
            Whole Mouse Genome G4122A A_52_P33841 AK035341.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_52_P33841 AK088930.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_52_P33841 AK035025.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_52_P33841 BC005560.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_52_P33841 X07967.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P514351 AK035341.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P514351 AK088930.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P514351 X07967.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P514351 NM_011725.3 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P514351 AK012549.1 mouse_transcriptome
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118419 - -
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115427 - -
            Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124964 - -
            Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09297577 - -
            Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03676512 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15095627 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15095626 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32389003 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388996 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388983 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388988 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388982 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388979 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388976 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32389002 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388998 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388997 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388986 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388991 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388994 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388977 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388978 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32389000 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388987 - -
            Agilent Six_ChIP_03_of_03_v2 A_68_P14549607 - -
            Agilent Six_ChIP_03_of_03_v2 A_68_P32389093 - -
            Agilent Six_ChIP_03_of_03_v2 A_68_P14549611 - -
            Agilent Six_ChIP_03_of_03_v2 A_68_P32389094 - -
            Array Name Probe IDs Targets Target Type
            Affymetrix Mouse Gene 1.0 ST Array 10604659 - -
            Affymetrix Mouse Gene 1.0 ST Array 10604660 - -
            Affymetrix Mouse Gene 1.0 ST Array 10604658 - -
            Affymetrix Mouse Gene 1.1 ST Array 10604656 - -
            Mouse Genome 430 2.0 Array 1422011_s_at - -
            Mouse Genome 430 2.0 Array 1422011_PM_s_at - -
            Affymetrix Mouse Clariom S Array PSR0X00014281.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014281.mm.1 TC0X00002247.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0X00014276.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014276.mm.1 TC0X00002247.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0X00014278.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014278.mm.1 TC0X00002247.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0X00014282.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014282.mm.1 TC0X00002247.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0X00014279.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014279.mm.1 TC0X00002247.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0X00014277.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014277.mm.1 TC0X00002247.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0X00014280.mm.1 NM_011725.4 Refseq
            Affymetrix Mouse Clariom S Array PSR0X00014280.mm.1 TC0X00002247.mm.1 MTA TC
            Mouse Expression Set 430 1422011_s_at - -
            Mouse Expression Set 430 1422011_PM_s_at - -
            Murine Genome U74v2 Set 92665_PM_f_at - -
            Murine Genome U74v2 Set 92665_f_at - -
            Mouse Genome 430A 2.0 Array 1422011_s_at - -
            Mouse Genome 430A 2.0 Array 1422011_PM_s_at - -
            Array Name Probe IDs Targets Target Type
            ABI Mouse Genome Survey Microarray 579959 - -
            ABI Mouse Genome Survey Microarray 886025 - -
            Array Name Probe IDs Targets Target Type
            MouseWG-6 v2 Expression BeadChip 5390091 - -
            Illumina mouse-6 v1.0 expression beadchip 5390091 - -
            Illumina mouseRef-8 v1.0 expression beadchip 0005390091 - -
            Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218723 - -
            Illumina mouse-6 v1.1 expression beadchip ILMN_1218723 - -
            mouse-6 v1.1 (Illumina) scl0022441.1_305-S - -
            Illumina Sentrix mouse-6.v1 bead array 5390091 - -
            Illumina Sentrix MouseRef-8 Expression BeadChip scl0022441.1_305-S - -
            Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390091 - -
            MouseRef-8 v2 Expression BeadChip ILMN_1218723 - -
            Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218723 - -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-g86sr:80/100.66.72.8:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0