Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27674871 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27674870 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2169621 |
AK077280.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2169621 |
AK008574.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2169621 |
BC033930.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2169621 |
BC115900.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2169621 |
BG092025.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2102574 |
AK077280.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2102574 |
AK085965.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2102574 |
BC033930.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2102574 |
BG092025.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2102574 |
NM_001163454.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562909 |
AK004264.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562909 |
AK135523.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562909 |
BC039885.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562909 |
BC052186.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562909 |
BC060276.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562909 |
NM_001081185.1 |
refseq_mrna_mouse_NM |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158889 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2102574 |
AK077280.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2102574 |
AK085965.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2102574 |
BC033930.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2102574 |
BG092025.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2102574 |
NM_001163454.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2169621 |
AK077280.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2169621 |
AK008574.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2169621 |
BC033930.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2169621 |
BC115900.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2169621 |
BG092025.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674898 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674911 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674903 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674888 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674857 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674891 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674868 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674902 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674853 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674907 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674864 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674852 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674862 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674883 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674858 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674890 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674869 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674881 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674879 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674887 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674871 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674859 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674901 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674880 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674855 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674872 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674860 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674900 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674875 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674885 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674897 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674906 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674874 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674865 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674867 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674878 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27674909 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P148254 |
AK041728.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P406021 |
AK085965.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P406021 |
NM_001163454.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P406021 |
NM_026465.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P304707 |
AK077280.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P304707 |
AK085965.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P304707 |
AK008574.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P304707 |
BC033930.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P304707 |
BC115900.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P114734 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09133051 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09133057 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09417174 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09253894 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27674871 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27674870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27674859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674888 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674858 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674891 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674892 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674862 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674868 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674871 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674881 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674865 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674890 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674876 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674887 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674869 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674872 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09054899 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674851 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09538122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674885 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674867 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674874 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674853 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27674884 |
- |
- |